Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124359.4 | NP_199792.1 | 9 - 10 | 1917 | 96 | |
GenBank mRNA | AY094459.1 | - | 9 - 10 | 1867 | 96 | |
BT002664.1 | - | 9 - 10 | 1792 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |