Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cpHsc70-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124369.4 | NP_199802.1 | 1 - 2 | 716 | 88 | |
GenBank mRNA | BT000919.1 | - | 1 - 2 | 580 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |