Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRO8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344882.1 | NP_001331201.1 | 5 - 6 | 1413 | 154 | |
NM_124395.5 | NP_199827.2 | 6 - 7 | 1328 | 154 | ||
GenBank mRNA | AY074287.1 | - | 6 - 7 | 1321 | 154 | |
BT003007.1 | - | 6 - 7 | 1260 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |