Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAP27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344892.1 | NP_001330206.1 | 6 - 7 | 887 | 70 | |
NM_124422.4 | NP_199851.2 | 6 - 7 | 962 | 70 | ||
GenBank mRNA | AY842027.1 | - | 6 - 7 | 672 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |