Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124471.4 | NP_568746.1 | 5 - 6 | 1459 | 83 | |
GenBank mRNA | AK227173.1 | - | 5 - 6 | 1152 | 83 | |
AY102125.1 | - | 5 - 6 | 1124 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |