Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124481.4 | NP_199915.1 | 6 - 7 | 614 | 77 | |
GenBank mRNA | AB120415.1 | - | 6 - 7 | 551 | 77 | |
AF370196.1 | - | 6 - 7 | 554 | 77 | ||
AY113932.1 | - | 6 - 7 | 474 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |