Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344918.1 | NP_001330683.1 | 2 - 3 | 176 | 61 | |
NM_001344919.1 | NP_001330687.1 | 2 - 3 | 176 | 61 | ||
NM_001344920.1 | NP_001330686.1 | 2 - 3 | 182 | 61 | ||
NM_124489.3 | NP_199923.1 | 2 - 3 | 214 | 61 | ||
GenBank mRNA | AK228538.1 | - | 2 - 3 | 121 | 61 | |
AY085077.1 | - | 2 - 3 | 147 | 61 | ||
BT004073.1 | - | 2 - 3 | 121 | 61 | ||
BT005116.1 | - | 2 - 3 | 110 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |