Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SWI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344934.1 | NP_001332209.1 | 6 - 7 | 1500 | 85 | |
NM_124512.3 | NP_568757.2 | 7 - 8 | 1502 | 85 | ||
GenBank mRNA | AF362001.1 | - | 6 - 7 | 1459 | 85 | |
AF466153.1 | - | 7 - 8 | 1501 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |