Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124589.3 | NP_200023.1 | 9 - 10 | 1019 | 68 | |
GenBank mRNA | BT030092.1 | - | 9 - 10 | 864 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |