Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PP2-A14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124591.6 | NP_200025.1 | 2 - 3 | 781 | 81 | |
GenBank mRNA | AY072088.1 | - | 2 - 3 | 617 | 81 | |
AY096374.1 | - | 2 - 3 | 450 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Arabidopsis thaliana Probe spans exons |