Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344986.1 | NP_001318786.1 | 3 - 4 | 518 | 153 | |
NM_001344987.1 | NP_001332093.1 | 3 - 4 | 540 | 153 | ||
NM_124596.2 | NP_200030.1 | 2 - 3 | 341 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |