Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344986.1 | NP_001318786.1 | 6 - 7 | 1326 | 104 | |
NM_001344987.1 | NP_001332093.1 | 6 - 7 | 1348 | 104 | ||
NM_124596.2 | NP_200030.1 | 5 - 6 | 1149 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |