Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBD13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203591.2 | NP_001190520.1 | 6 - 7 | 992 | 143 | |
NM_001344990.1 | NP_001331264.1 | 6 - 7 | 879 | 143 | ||
NM_124602.4 | NP_200036.1 | 5 - 6 | 1095 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |