Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXO70A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124613.4 | NP_200047.3 | 4 - 5 | 845 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |