Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124635.4 | NP_568775.1 | 16 - 17 | 1765 | 69 | |
GenBank mRNA | AF360236.1 | - | 16 - 17 | 1628 | 69 | |
AY040035.1 | - | 15 - 16 | 1442 | 69 | ||
DQ324529.1 | - | 15 - 16 | 1442 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |