Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATIM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345015.1 | NP_001330129.1 | 10 - 11 | 2348 | 127 | |
NM_124669.3 | NP_200103.1 | 10 - 11 | 2204 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |