Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAP46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124679.3 | NP_568783.1 | 6 - 7 | 777 | 115 | |
GenBank mRNA | AF133708.1 | - | 6 - 7 | 720 | 115 | |
BT004072.1 | - | 6 - 7 | 721 | 115 | ||
BT006150.1 | - | 6 - 7 | 604 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |