Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAP46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124679.3 | NP_568783.1 | 7 - 8 | 857 | 76 | |
GenBank mRNA | AF133708.1 | - | 7 - 8 | 800 | 76 | |
BT004072.1 | - | 7 - 8 | 801 | 76 | ||
BT006150.1 | - | 7 - 8 | 684 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |