Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAP46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124679.3 | NP_568783.1 | 8 - 9 | 1137 | 66 | |
GenBank mRNA | AF133708.1 | - | 8 - 9 | 1080 | 66 | |
BT004072.1 | - | 8 - 9 | 1081 | 66 | ||
BT006150.1 | - | 8 - 9 | 964 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |