Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345042.1 | NP_001332167.1 | 3 - 4 | 522 | 63 | |
NM_124697.5 | NP_200130.1 | 3 - 4 | 530 | 63 | ||
GenBank mRNA | BT015760.1 | - | 3 - 4 | 456 | 63 | |
BT020199.1 | - | 2 - 3 | 393 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Arabidopsis thaliana Probe spans exons |