Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085281.2 | NP_001078750.1 | 7 - 8 | 821 | 109 | |
NM_001345040.1 | NP_001332166.1 | 6 - 7 | 973 | 109 | ||
NM_001345041.1 | NP_001332168.1 | 7 - 8 | 826 | 109 | ||
NM_001345042.1 | NP_001332167.1 | 6 - 7 | 783 | 109 | ||
NM_001345043.1 | NP_001332169.1 | 3 - 4 | 676 | 109 | ||
NM_124697.5 | NP_200130.1 | 6 - 7 | 763 | 109 | ||
GenBank mRNA | AK228857.1 | - | 7 - 8 | 821 | 109 | |
BT015760.1 | - | 6 - 7 | 689 | 109 | ||
BT020199.1 | - | 5 - 6 | 626 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |