Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085281.2 | NP_001078750.1 | 8 - 9 | 883 | 68 | |
NM_001345040.1 | NP_001332166.1 | 7 - 8 | 1035 | 68 | ||
NM_001345041.1 | NP_001332168.1 | 8 - 9 | 888 | 68 | ||
NM_001345042.1 | NP_001332167.1 | 7 - 8 | 845 | 68 | ||
NM_001345043.1 | NP_001332169.1 | 4 - 5 | 738 | 68 | ||
NM_124697.5 | NP_200130.1 | 7 - 8 | 825 | 68 | ||
GenBank mRNA | AK228857.1 | - | 8 - 9 | 883 | 68 | |
BT015760.1 | - | 7 - 8 | 751 | 68 | ||
BT020199.1 | - | 6 - 7 | 688 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |