Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085281.2 | NP_001078750.1 | 9 - 10 | 979 | 143 | |
NM_001345040.1 | NP_001332166.1 | 8 - 9 | 1131 | 143 | ||
NM_001345041.1 | NP_001332168.1 | 9 - 10 | 984 | 143 | ||
NM_001345042.1 | NP_001332167.1 | 8 - 9 | 941 | 143 | ||
NM_001345043.1 | NP_001332169.1 | 5 - 6 | 834 | 143 | ||
NM_124697.5 | NP_200130.1 | 8 - 9 | 921 | 143 | ||
GenBank mRNA | AK228857.1 | - | 9 - 10 | 979 | 143 | |
BT015760.1 | - | 8 - 9 | 847 | 143 | ||
BT020199.1 | - | 7 - 8 | 784 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |