Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDG29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124722.3 | NP_200155.2 | 6 - 7 | 1841 | 94 | |
GenBank mRNA | AK117221.1 | - | 6 - 7 | 1841 | 94 | |
BT005981.1 | - | 6 - 7 | 1364 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |