Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDG29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124722.3 | NP_200155.2 | 8 - 9 | 2028 | 116 | |
GenBank mRNA | AK117221.1 | - | 8 - 9 | 2028 | 116 | |
BT005981.1 | - | 8 - 9 | 1551 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |