Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124726.4 | NP_200159.1 | 4 - 5 | 1071 | 76 | |
GenBank mRNA | AY059881.1 | - | 4 - 5 | 836 | 76 | |
AY093383.1 | - | 4 - 5 | 744 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |