Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAMI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345079.1 | NP_001331145.1 | 1 - 2 | 546 | 100 | |
NM_124791.3 | NP_568804.1 | 1 - 2 | 563 | 100 | ||
GenBank mRNA | BT025794.1 | - | 1 - 2 | 266 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |