Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ILL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124794.3 | NP_200225.1 | 2 - 3 | 613 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |