Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRXL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336562.1 | NP_001324041.1 | 5 - 6 | 984 | 89 | |
NM_001336563.1 | NP_001324042.1 | 3 - 4 | 1106 | 89 | ||
NM_129113.5 | NP_565809.1 | 5 - 6 | 1004 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |