Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scpl26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129136.5 | NP_181121.1 | 6 - 7 | 1206 | 109 | |
GenBank mRNA | AY088044.1 | - | 6 - 7 | 1026 | 109 | |
BT029178.1 | - | 6 - 7 | 1012 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |