Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E2F3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129160.4 | NP_565831.3 | 5 - 6 | 890 | 60 | |
GenBank mRNA | AF242582.1 | - | 5 - 6 | 1153 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |