Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APG10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129181.4 | NP_181165.1 | 5 - 6 | 743 | 62 | |
GenBank mRNA | AB006139.1 | - | 5 - 6 | 723 | 62 | |
AK118659.1 | - | 5 - 6 | 719 | 62 | ||
AY140058.1 | - | 5 - 6 | 719 | 62 | ||
BT002552.1 | - | 5 - 6 | 694 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |