Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336601.1 | NP_001324936.1 | 10 - 11 | 1742 | 68 | |
NM_129209.4 | NP_181192.1 | 10 - 11 | 1543 | 68 | ||
GenBank mRNA | AF424603.1 | - | 10 - 11 | 1273 | 68 | |
AY150418.1 | - | 10 - 11 | 1200 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |