Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336601.1 | NP_001324936.1 | 11 - 12 | 1793 | 65 | |
NM_129209.4 | NP_181192.1 | 11 - 12 | 1594 | 65 | ||
GenBank mRNA | AF424603.1 | - | 11 - 12 | 1324 | 65 | |
AY150418.1 | - | 11 - 12 | 1251 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |