Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT73C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129230.3 | NP_181213.1 | 1 | 685 | 92 | |
GenBank mRNA | BT026383.1 | - | 1 | 627 | 92 | |
KJ138870.1 | - | 1 | 627 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |