Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336618.1 | NP_001323430.1 | 13 - 14 | 3417 | 86 | |
NM_129257.4 | NP_181238.4 | 13 - 14 | 3417 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |