Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084551.1 | NP_001078020.1 | 6 - 7 | 1582 | 76 | |
NM_129359.4 | NP_565879.1 | 6 - 7 | 1583 | 76 | ||
GenBank mRNA | AF359240.1 | - | 5 - 6 | 1490 | 76 | |
AY048248.1 | - | 6 - 7 | 1557 | 76 | ||
AY139799.1 | - | 5 - 6 | 1474 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |