Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSRP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129370.5 | NP_181349.1 | 2 - 3 | 327 | 80 | |
GenBank mRNA | AF236826.1 | - | 2 - 3 | 246 | 80 | |
AY052215.1 | - | 2 - 3 | 269 | 80 | ||
AY060491.1 | - | 2 - 3 | 227 | 80 | ||
AY088244.1 | - | 2 - 3 | 292 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |