Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFIIS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129413.4 | NP_181390.1 | 1 - 2 | 424 | 77 | |
GenBank mRNA | AK117557.1 | - | 1 - 2 | 328 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |