Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATATH8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129476.3 | NP_565900.1 | 2 - 3 | 678 | 63 | |
NM_129477.1 | NP_565901.1 | 2 - 3 | 678 | 63 | ||
GenBank mRNA | AY048299.1 | - | 2 - 3 | 678 | 63 | |
BT002715.1 | - | 2 - 3 | 506 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |