Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PnsL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036437.3 | NP_001031514.2 | 5 - 6 | 535 | 93 | |
NM_001084553.1 | NP_001078022.1 | 5 - 6 | 535 | 93 | ||
NM_129505.4 | NP_565906.1 | 5 - 6 | 577 | 93 | ||
GenBank mRNA | AY039968.1 | - | 5 - 6 | 473 | 93 | |
AY085632.1 | - | 5 - 6 | 468 | 93 | ||
AY133798.1 | - | 5 - 6 | 440 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |