Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129534.4 | NP_030522.1 | 3 - 4 | 1218 | 89 | |
GenBank mRNA | AY128315.1 | - | 3 - 4 | 1142 | 89 | |
BT000044.1 | - | 3 - 4 | 1014 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Arabidopsis thaliana Probe spans exons |