Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125006.1 | NP_001118478.1 | 5 - 6 | 582 | 141 | |
NM_129536.5 | NP_030524.1 | 7 - 8 | 783 | 141 | ||
GenBank mRNA | BT025792.1 | - | 7 - 8 | 644 | 141 | |
M98336.1 | - | 7 - 8 | 659 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |