Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336807.1 | NP_001324597.1 | 4 - 5 | 812 | 81 | |
NM_001336808.1 | NP_001324596.1 | 4 - 5 | 812 | 81 | ||
NM_129551.4 | NP_181522.1 | 4 - 5 | 984 | 81 | ||
GenBank mRNA | BT000443.1 | - | 4 - 5 | 788 | 81 | |
BT010348.1 | - | 4 - 5 | 757 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |