Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336807.1 | NP_001324597.1 | 7 - 8 | 1158 | 140 | |
NM_001336808.1 | NP_001324596.1 | 7 - 8 | 1158 | 140 | ||
NM_129551.4 | NP_181522.1 | 7 - 8 | 1330 | 140 | ||
GenBank mRNA | BT000443.1 | - | 7 - 8 | 1134 | 140 | |
BT010348.1 | - | 7 - 8 | 1103 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |