Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRPD1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336818.1 | NP_001323985.1 | 5 - 6 | 645 | 77 | |
NM_129561.4 | NP_181532.2 | 5 - 6 | 645 | 77 | ||
GenBank mRNA | AY826516.1 | - | 4 - 5 | 371 | 77 | |
AY927744.1 | - | 4 - 5 | 371 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |