Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129597.4 | NP_181568.1 | 5 - 6 | 1798 | 79 | |
GenBank mRNA | AF506030.1 | - | 5 - 6 | 1625 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |