Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129617.5 | NP_181587.2 | 6 - 7 | 969 | 113 | |
GenBank mRNA | BT022032.1 | - | 6 - 7 | 678 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |