Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336876.1 | NP_001323724.1 | 10 - 11 | 2081 | 94 | |
NM_129656.4 | NP_565944.1 | 10 - 11 | 2140 | 94 | ||
GenBank mRNA | AF048705.1 | - | 10 - 11 | 2045 | 94 | |
AY099701.1 | - | 10 - 11 | 2036 | 94 | ||
BT000300.1 | - | 10 - 11 | 1804 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |