Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129693.4 | NP_181661.1 | 2 - 3 | 812 | 77 | |
GenBank mRNA | BT015732.1 | - | 2 - 3 | 578 | 77 | |
BT020191.1 | - | 2 - 3 | 537 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |